Order Entry
Sweden
ContactUsLinkComponent
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain B/Tokio/53/99. The Influenza Virus was puri fied by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81790-50
Supplier:  Biorbyt
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain B/Tokio/53/99. The Influenza Virus was puri fied by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81790-50
Supplier:  Biorbyt
Related Information