0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Human C4c produced in Human plasma having a molecular mass of 210 kDa. 1 * 200 µG
Catalog # BIRBORB81044-200
Supplier:
Biorbyt
Expand image
Human C4c produced in Human plasma having a molecular mass of 210 kDa. 1 * 200 µG
Catalog # BIRBORB81044-200
Supplier:
Biorbyt
Related Information
Documentation
Datasheet
orb81044
Documentation