Order Entry
Sweden
ContactUsLinkComponent
BIN1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 459 amino acids (1-439 a.a.) an d having a molecular mass of 50.4 kDa. The BIN1 is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 20 µG
Catalog # BIRBORB80966-20
Supplier:  Biorbyt
BIN1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 459 amino acids (1-439 a.a.) an d having a molecular mass of 50.4 kDa. The BIN1 is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 20 µG
Catalog # BIRBORB80966-20
Supplier:  Biorbyt