Order Entry
Sweden
ContactUsLinkComponent
Stratifin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 248 amino acids (1-248) a nd having a molecular mass of 27.7 kDa. Stratifin is purified by proprietary chromatographic techniques. 1 * 5 µG
  BIRBORB80885-5
 :  Biorbyt
Stratifin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 248 amino acids (1-248) a nd having a molecular mass of 27.7 kDa. Stratifin is purified by proprietary chromatographic techniques. 1 * 5 µG
  BIRBORB80885-5
 :  Biorbyt