Order Entry
Sweden
ContactUsLinkComponent
Glucagon Human Synthetic peptide is a single, non-glycosylated, polypeptide chain containing 29 amino acids and having a molecular m ass of 3485 Dalton.The Glucagon is purified by proprietary chromatographic techniques. 1 * 4 mg
Catalog # BIRBORB80725-4
Supplier:  Biorbyt
Glucagon Human Synthetic peptide is a single, non-glycosylated, polypeptide chain containing 29 amino acids and having a molecular m ass of 3485 Dalton.The Glucagon is purified by proprietary chromatographic techniques. 1 * 4 mg
Catalog # BIRBORB80725-4
Supplier:  Biorbyt
Related Information