Order Entry
Sweden
ContactUsLinkComponent
LGALS1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids and having a molecular mass of 14.5 kDa.The LGALS1 is purified by proprietary chromatographic techniques. 1 * 50 µG
  BIRBORB80328-50
 :  Biorbyt
LGALS1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids and having a molecular mass of 14.5 kDa.The LGALS1 is purified by proprietary chromatographic techniques. 1 * 50 µG
  BIRBORB80328-50
 :  Biorbyt