Order Entry
Sweden
ContactUsLinkComponent
Melanotan-I has the amino acid sequence of Ac-Ser-Tyr-Ser-Nle-Glu-His-D-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2 and molecular weight of 1645 .82 Dalton. 1 * 20 mg
Catalog # BIRBORB80742-20
Supplier:  Biorbyt
Melanotan-I has the amino acid sequence of Ac-Ser-Tyr-Ser-Nle-Glu-His-D-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2 and molecular weight of 1645 .82 Dalton. 1 * 20 mg
Catalog # BIRBORB80742-20
Supplier:  Biorbyt