Order Entry
Sweden
ContactUsLinkComponent
Keratinocyte Growth Factor-2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 170 ami no acids and having a molecular mass of 19300 Dalton. Keratinocyte Growth Factor is highly related to KGF-1( 1 * 5 µG
Catalog # BIRBORB80484-5
Supplier:  Biorbyt
Keratinocyte Growth Factor-2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 170 ami no acids and having a molecular mass of 19300 Dalton. Keratinocyte Growth Factor is highly related to KGF-1( 1 * 5 µG
Catalog # BIRBORB80484-5
Supplier:  Biorbyt
Related Information