Order Entry
Sweden
ContactUsLinkComponent
Lanreotide is peptide inhibitor of number of endocrine, neuroendocrine, exocrine and paracrine functions. It shows good affinity for peripheral somatostatin receptors (anterior pituitary and pancreatic). In contrast, its affinity for centra 1 * 200 µG
  BIRBORB80736-200
 :  Biorbyt
Lanreotide is peptide inhibitor of number of endocrine, neuroendocrine, exocrine and paracrine functions. It shows good affinity for peripheral somatostatin receptors (anterior pituitary and pancreatic). In contrast, its affinity for centra 1 * 200 µG
  BIRBORB80736-200
 :  Biorbyt
Frequently Bought Together