Order Entry
Sweden
ContactUsLinkComponent
Lanreotide is peptide inhibitor of number of endocrine, neuroendocrine, exocrine and paracrine functions. It shows good affinity for peripheral somatostatin receptors (anterior pituitary and pancreatic). In contrast, its affinity for centra 1 * 1 mg
Catalog # BIRBORB80736-1
Supplier:  Biorbyt
Lanreotide is peptide inhibitor of number of endocrine, neuroendocrine, exocrine and paracrine functions. It shows good affinity for peripheral somatostatin receptors (anterior pituitary and pancreatic). In contrast, its affinity for centra 1 * 1 mg
Catalog # BIRBORB80736-1
Supplier:  Biorbyt