Order Entry
Sweden
ContactUsLinkComponent
Human TNFSF4/OX40L ELISA Kit. OX40L is the ligand for CD134 and is expressed on such cells as DC2s enabling amplification of Th2 cel l differentiation. OX40L has also been designated CD252. By analysis of an interspecific backcross, the mous 1 * 96 items
Catalog # BIRBORB138070-96
Supplier:  Biorbyt
Human TNFSF4/OX40L ELISA Kit. OX40L is the ligand for CD134 and is expressed on such cells as DC2s enabling amplification of Th2 cel l differentiation. OX40L has also been designated CD252. By analysis of an interspecific backcross, the mous 1 * 96 items
Catalog # BIRBORB138070-96
Supplier:  Biorbyt