FLAG TAG ANTIBODY 1 * 200 µG
Catalog # BIRBORB129610-200
Supplier: Biorbyt
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Förseglingsfilm för manuell applicering, Parafilm® M
Supplier: HEATHROW
VWR®, Transferpipetter, hög prestanda
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandskar
Supplier: VWR Collection
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade för HPLC
Supplier: VWR Chemicals
Vatten, HiPerSolv CHROMANORM® för HPLC
Supplier: VWR Chemicals
VWR®, Serologisk pipett, standard
Supplier: VWR Collection
Handskar, nitril, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrilhandskar
Supplier: VWR Collection
VWR®, Ansiktsservett
Supplier: VWR Collection
Teststrips, MQuant®, Supelco®
Supplier: Merck