Recombinant Rat Oncostatin-M (OSM) is a multifunctional cytokine that belongs to the Interleukin-6 subfamily. Among the family membe rs, OSM is most closely related to leukemia inhibitory factor (LIF) and it in fact utilizes the LIF receptor 1 * 2 µG
Catalog # BIRBORB107792-2
Supplier: Biorbyt
Frequently Bought Together