Order Entry
Sweden
ContactUsLinkComponent
Hi-sens hf+tds sample intro kit rad icp. high sensitivity, inert sample intro kit for hf/high tds with 7 series radial icp. incl ine rt sturman masters s/chamber, oneneb + fep transfer tube 1 * 1 items
Catalog # VARI9810046390
Supplier:  VARIAN
Hi-sens hf+tds sample intro kit rad icp. high sensitivity, inert sample intro kit for hf/high tds with 7 series radial icp. incl ine rt sturman masters s/chamber, oneneb + fep transfer tube 1 * 1 items
Catalog # VARI9810046390
Supplier:  VARIAN