0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
BUFFER SOLUTION PH 10,00 (A) 20C (CARBO 1 * 15 mL
FFCHFF64660015
:
FF CHEMICALS
BUFFER SOLUTION PH 10,00 (A) 20C (CARBO 1 * 15 mL
FFCHFF64660015
:
FF CHEMICALS