0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
BUFFER SOLUTION PH 9,00 (A) 20C (TRIS B 1 * 500 mL
FFCHFF64650500
:
FF CHEMICALS
BUFFER SOLUTION PH 9,00 (A) 20C (TRIS B 1 * 500 mL
FFCHFF64650500
:
FF CHEMICALS