0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Phenyl Hexylx 5µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-05N02S
Supplier:
Trajan Scientific and Medical
Expand image
Phenyl Hexylx 5µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-05N02S
Supplier:
Trajan Scientific and Medical