Order Entry
Sweden
ContactUsLinkComponent
C8H x 5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
  SGEA2C83-05M02P
C8H x 5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
  SGEA2C83-05M02P