0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C18G x 3µm particle size x 120Å pore size, 50mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-03L46S
Supplier:
Trajan Scientific and Medical
Expand image
C18G x 3µm particle size x 120Å pore size, 50mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-03L46S
Supplier:
Trajan Scientific and Medical