Data Logger, Einstein LabMate
Data logger for science and education, einstein™ LabMate+
Catalog # FOURENLMSEN
Supplier: FOURIER SYSTEMS
Jump to:
Product Details & Documents
The LabMate + is a science, technology engineering and mathematics education tool, ideal for primary and secondary schools already equipped with tablets or computers.
- Small and durable enough to put in a terrarium, or take in the pocket for outdoor science exploration
- Range of six built-in sensors plus the connectivity with four inputs for a range of optional external sensors (eight sensors possible with splitters)
- Runs experiment over 24 hours without charging
Pairs to any device via Bluetooth or a USB cable. Built-in sensors include: Heart Rate Sensor, Ambient Humidity Sensor, Light Sensor, UV Sensor, Pressure Sensor, Ambient Temperature.
: Pressure kit must be ordered separately.
Specifications
- Description:Data logger for science and education, einstein™ LabMate+
- Cat. No.:FOURENLMSEN
- Interface:USB
Accessories
Product Information
- Catalog #AvailabilityPrice
- Sensor adapter
Product Information
- Catalog #AvailabilityPrice
- Blood pressure sensor for einstein™ LabMate
Product Information
- Catalog #AvailabilityPrice
- Sensor cable