BUFFER SOLUTION PH 7,00 (A) 20C (PHOSPH 1 * 6 Items
Catalog # FFCHFF64576015
Supplier: FF CHEMICALS
Frequently Bought Together
Handskar, nitril, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Serologisk pipett, standard
Supplier: VWR Collection
Förseglingsfilm för manuell applicering, Parafilm® M
Supplier: HEATHROW
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade för HPLC
Supplier: VWR Chemicals
Vatten, HiPerSolv CHROMANORM® för HPLC
Supplier: VWR Chemicals
VWR®, Transferpipetter, hög prestanda
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandskar
Supplier: VWR Collection
VWR®, Nitrilhandskar
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Ansiktsservett
Supplier: VWR Collection
Teststrips, MQuant®, Supelco®
Supplier: Merck