Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLUTION PH 10,00 20C (BORIC 1 * 6 Items
Catalog # FFCHFF1826015
Supplier:
FF CHEMICALS
Expand image
BUFFER SOLUTION PH 10,00 20C (BORIC 1 * 6 Items
Catalog # FFCHFF1826015
Supplier:
FF CHEMICALS