Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLUTION PH 10,00 (A) 20C (BORIC 1 * 500 mL
Catalog # FFCHFF64580500
Supplier:
FF CHEMICALS
Expand image
BUFFER SOLUTION PH 10,00 (A) 20C (BORIC 1 * 500 mL
Catalog # FFCHFF64580500
Supplier:
FF CHEMICALS