0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
BUFFER SOLUTION PH 7,00 (A) 20C (PHOSPH 1 * 15 mL
FFCHFF64570015
:
FF CHEMICALS
BUFFER SOLUTION PH 7,00 (A) 20C (PHOSPH 1 * 15 mL
FFCHFF64570015
:
FF CHEMICALS