Reagent containers for Microlab® STAR line
Reagent container without lid, non self-standing, 1T, Non self-standing, 1T, 120 ml, 155,9×21,1×91,5 mm, Transparent, Individually wrapped
Katalog # 736-0833
Marketsource
Leverantör: HAMILTON ROBOTICS
Reagent containers for Microlab® STAR line
Katalog # 736-0833
Leverantör: HAMILTON ROBOTICS
Marketsource
Gå till:
Product Details & Documents
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
- Special bottom geometry minimises residual volumes
- With volume markings, without or with lid
- With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
- IVD compliant
- Autoclavable
Ordering information: The carriers are not included and need to be ordered separately.
Documents
Certificates
Sök efter LOT-nummerSpecifikationer
- Packed:Individually wrapped
- Type:Non self-standing, 1T
- Capacity:120 ml
- Colour:Transparent
- Description:Reagent container without lid, non self-standing, 1T
- Seq:08
- W×D×H:155,9×21,1×91,5 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



