0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Metal ferrules, Brass, nickel-plated to fit ASTM thermometers Nr. 9 C/9 F and 10 C/ 10 F 1 * 1 items
Catalog # AMARA312004
Supplier:
Amarell
Expand image
Metal ferrules, Brass, nickel-plated to fit ASTM thermometers Nr. 9 C/9 F and 10 C/ 10 F 1 * 1 items
Catalog # AMARA312004
Supplier:
Amarell