Order Entry
Sweden
ContactUsLinkComponent
MGC11256 (Human) Recombinant protein (Q01). Human MGC11256 partial ORF ( NP_077300, 162 A.A. - 258 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00079174-Q012
Supplier:  Abnova
MGC11256 (Human) Recombinant protein (Q01). Human MGC11256 partial ORF ( NP_077300, 162 A.A. - 258 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00079174-Q012
Supplier:  Abnova