Order Entry
Sweden
ContactUsLinkComponent
IGHV4-31 (Human) Recombinant protein (P02). Human IGHV4-31 full-length ORF ( AAH53984.1, 1 A.A. - 470 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00028396-P022
Supplier:  Abnova
IGHV4-31 (Human) Recombinant protein (P02). Human IGHV4-31 full-length ORF ( AAH53984.1, 1 A.A. - 470 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00028396-P022
Supplier:  Abnova
Frequently Bought Together