Order Entry
Sweden
ContactUsLinkComponent
KIAA0527 (Human) Recombinant protein (Q01). Human KIAA0527 partial ORF ( XP_171054, 284 A.A. - 383 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00026032-Q012
Supplier:  Abnova
KIAA0527 (Human) Recombinant protein (Q01). Human KIAA0527 partial ORF ( XP_171054, 284 A.A. - 383 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00026032-Q012
Supplier:  Abnova