Order Entry
Sweden
ContactUsLinkComponent
CDC42EP2 (Human) Recombinant protein (Q01). Human CDC42EP2 partial ORF ( NP_006770, 102 A.A. - 210 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00010435-Q012
Supplier:  Abnova
CDC42EP2 (Human) Recombinant protein (Q01). Human CDC42EP2 partial ORF ( NP_006770, 102 A.A. - 210 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00010435-Q012
Supplier:  Abnova