Order Entry
Sweden
ContactUsLinkComponent
D8S2298E (Human) Recombinant protein (Q01). Human D8S2298E partial ORF ( NP_005662, 173 A.A. - 268 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00007993-Q012
Supplier:  Abnova
D8S2298E (Human) Recombinant protein (Q01). Human D8S2298E partial ORF ( NP_005662, 173 A.A. - 268 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00007993-Q012
Supplier:  Abnova