Order Entry
Sweden
ContactUsLinkComponent
CDC42BPA (Human) Recombinant protein (Q01). Human CDC42BPA partial ORF ( NP_003598, 551 A.A. - 650 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00008476-Q012
Supplier:  Abnova
CDC42BPA (Human) Recombinant protein (Q01). Human CDC42BPA partial ORF ( NP_003598, 551 A.A. - 650 A.A.) Recombinant protein with GS T-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00008476-Q012
Supplier:  Abnova