Y-27632, Millipore®
100 mg
Katalog # 688000-100MG
Leverantör: Merck Millipore (Calbiochem)
Y-27632, Millipore®
Katalog # 688000-100MG
Leverantör: Merck Millipore (Calbiochem)
CAS-nummer: 146986-50-7
Gå till:
Product Details & Documents
A highly potent, cell-permeable, reversible, and selective inhibitor of Rho-associated protein kinases (Ki = 140 nm for p160ROCK). Also inhibits ROCK-II with equal potency. Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contraction.
- Inhibition is competitive with respect to ATP
- Exhibits 10- to 50-fold lower affinity for PKCε than p160ROCK
- Pretreatment with Y-27632 reduces the risk of lung injury in rat models of sepsis
- Does not affect the activity of p21-activated protein kinase (PAK) even at higher concentrations (~100 µm)
Prevents apoptosis and enhances the survival and cloning efficiency of dissociated hES cells without affecting their pluripotency. A 5 mm (500 µg/296 µl) solution of Y-27632 in H2O and a 10 mm solution in DMSO are also available.
Synonymer: trans-4-((R)-1-Aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide
- CAS-nummer:146986-50-7
- Formel:C₁₄H₂₁N₃O
- MDL-nummer:MFCD03093855
- Molecular Weight:247.34 g/mol
Specifikationer
Specifikationer
- Pk:100 mg



