Order Entry
Sweden
ContactUsLinkComponent
Color Coding Cap Inserts for Cryo Tubes. Polypropylene. For cryotubes 1.2 to 5.0 ml. Color Red. 1 * 1.000 items
  RATI5110002
 :  RATIOLAB
Color Coding Cap Inserts for Cryo Tubes. Polypropylene. For cryotubes 1.2 to 5.0 ml. Color Red. 1 * 1.000 items
  RATI5110002
 :  RATIOLAB