Order Entry
Sweden
ContactUsLinkComponent
LID FLAT FLANGE 4 NECKS Lids LF 60, 2 x 19/26 angled side necks and 1 x 19/26 straight side neck with a 29/32 central neck, like 6.1041.41 Lenz ref. no.: 81002180 1 * 1 items
Catalog # LENZ81002180
LID FLAT FLANGE 4 NECKS Lids LF 60, 2 x 19/26 angled side necks and 1 x 19/26 straight side neck with a 29/32 central neck, like 6.1041.41 Lenz ref. no.: 81002180 1 * 1 items
Catalog # LENZ81002180