Order Entry
Sweden
Orders LinkContactUsLinkComponent
Buffer solutions, ready to use, referenced at 25 °C, Certipur®

Buffer solutions, ready to use, referenced at 25 °C, Certipur®

1,00, Glycine, sodium chloride, hydrogen chloride, 500 mL
Katalog # 1.09441.0500
Leverantör:   Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Katalog # 1.09441.0500
Leverantör:  Merck
CAS-nummer:  
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Buffer solutions, ready to use, referenced at 25 °C, Certipur®

1,00, Glycine, sodium chloride, hydrogen chloride, 500 mL

CAS-nummer:  
Price
625,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 500 mL
Product Details & Documents

Materials for the precise calibration and monitoring of pH meters.

  • Traceable to NIST primary standard and PTB - deviation max. ±0,01 (except pH 10 with ±0,02)
  • Larger 4 litre pack sizes in easy to use Titripac® packaging which eliminates contamination issues
  • Colour coded buffers at pH values 4,00, 7,00 and 10,00 are available
  • Reference temperature: +25 °C

All buffer solutions are measured and qualified in a DIN EN ISO 17025/IEC accredited laboratory for pH measurement.

Packaging: Plastic bottles (500 ml) or PE/aluminium Titripac® pack (4 litre)

Specifikationer

Specifikationer

  • Pk:
    500 mL
  • pH value:
    1,00
  • Contents:
    Glycine, sodium chloride, hydrogen chloride

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.