Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, tyro3 (human) recombinant protein, human tyro3 (np_006284.2, 453 a.a. - 890 a.a.) partial recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5809
Supplier:  Abnova
recombinant/native protein, tyro3 (human) recombinant protein, human tyro3 (np_006284.2, 453 a.a. - 890 a.a.) partial recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5809
Supplier:  Abnova
Related Information