Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, pkn1 (human) recombinant protein, human pkn1 (np_002732.3, 1 a.a. - 942 a.a.) full-length recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5795
Supplier:  Abnova
recombinant/native protein, pkn1 (human) recombinant protein, human pkn1 (np_002732.3, 1 a.a. - 942 a.a.) full-length recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5795
Supplier:  Abnova