Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, pak1 (human) recombinant protein, human pak1 (np_002567.3, 1 a.a. - 545 a.a.) full-length recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5788
 :  Abnova
recombinant/native protein, pak1 (human) recombinant protein, human pak1 (np_002567.3, 1 a.a. - 545 a.a.) full-length recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5788
 :  Abnova
Frequently Bought Together