0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
recombinant/native protein, flt1 (human) recombinant protein, human flt1 (np_002010.1, 781 a.a. - 1338 a.a.) partial recombinant pro tein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
ABNOP5768
:
Abnova
recombinant/native protein, flt1 (human) recombinant protein, human flt1 (np_002010.1, 781 a.a. - 1338 a.a.) partial recombinant pro tein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
ABNOP5768
:
Abnova
TDS_ABNOP5768.pdf