Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, fgfr4 (v550l) (human) recombinant protein, human fgfr4 (np_002002.3, 460 a.a. - 802 a.a.) v550l mutant p artial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5759
 :  Abnova
recombinant/native protein, fgfr4 (v550l) (human) recombinant protein, human fgfr4 (np_002002.3, 460 a.a. - 802 a.a.) v550l mutant p artial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5759
 :  Abnova
Frequently Bought Together