Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, abl1 (t315i) (human) recombinant protein, human abl1 (np_005148.2, 2 a.a. - 1130 a.a.) t315i mutant full length recombinant protein with his tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5752
Supplier:  Abnova
recombinant/native protein, abl1 (t315i) (human) recombinant protein, human abl1 (np_005148.2, 2 a.a. - 1130 a.a.) t315i mutant full length recombinant protein with his tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5752
Supplier:  Abnova
Related Information