Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, abl1 (e255k) (human) recombinant protein, human abl1 (np_005148.2, 2 a.a. - 1130 a.a.) e255k mutant full length recombinant protein with his tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5740
 :  Abnova
recombinant/native protein, abl1 (e255k) (human) recombinant protein, human abl1 (np_005148.2, 2 a.a. - 1130 a.a.) e255k mutant full length recombinant protein with his tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5740
 :  Abnova
Frequently Bought Together