Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, pdgfra (d842v) (human) recombinant protein, human pdgfra (np_006197.1, 550 a.a. - 1089 a.a.) d842v mutan t partial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5739
Supplier:  Abnova
recombinant/native protein, pdgfra (d842v) (human) recombinant protein, human pdgfra (np_006197.1, 550 a.a. - 1089 a.a.) d842v mutan t partial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5739
Supplier:  Abnova