Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, met (human) recombinant protein, human met (np_000236.2, 956 a.a. - 1390 a.a.) partial recombinant prote in with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5728
Supplier:  Abnova
recombinant/native protein, met (human) recombinant protein, human met (np_000236.2, 956 a.a. - 1390 a.a.) partial recombinant prote in with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5728
Supplier:  Abnova
Related Information