Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, map3k14 (human) recombinant protein, human map3k14 (np_003945.1, 658 a.a. - 1114 a.a.) partial recombina nt protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5725
Supplier:  Abnova
recombinant/native protein, map3k14 (human) recombinant protein, human map3k14 (np_003945.1, 658 a.a. - 1114 a.a.) partial recombina nt protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5725
Supplier:  Abnova