Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, ikbke (human) recombinant protein, human ikbke (np_054721.1, 1 a.a. - 716 a.a.) full-length recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5721
Supplier:  Abnova
recombinant/native protein, ikbke (human) recombinant protein, human ikbke (np_054721.1, 1 a.a. - 716 a.a.) full-length recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5721
Supplier:  Abnova
Related Information