Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, epha5 (human) recombinant protein, human epha5 (np_004430.3, 662 a.a. - 948 a.a.) partial recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5706
Supplier:  Abnova
recombinant/native protein, epha5 (human) recombinant protein, human epha5 (np_004430.3, 662 a.a. - 948 a.a.) partial recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5706
Supplier:  Abnova