Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, egfr (t790m) (human) recombinant protein, human egfr (np_005219.2, 669 a.a. - 1210 a.a.) t790m mutant pa rial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5702
Supplier:  Abnova
recombinant/native protein, egfr (t790m) (human) recombinant protein, human egfr (np_005219.2, 669 a.a. - 1210 a.a.) t790m mutant pa rial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5702
Supplier:  Abnova
Related Information