Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, egfr (d746-750) (human) recombinant protein, human egfr (np_005219.2, 669 a.a. - 1210 a.a.) parial recom binant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5698
 :  Abnova
recombinant/native protein, egfr (d746-750) (human) recombinant protein, human egfr (np_005219.2, 669 a.a. - 1210 a.a.) parial recom binant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
  ABNOP5698
 :  Abnova
Frequently Bought Together